Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-PEG2-DBCO | 2639395-48-3 | MFCD34178807 | 95.4% | 722.83 g/mol | C44H42N4O6 | 10 MG
SDP

Catalog No. 5000286577
Encompass_Preferred

DBCO-PEG2-DBCO is a bifunctional polyethylene glycol (PEG) linker bearing two terminal dibenzocyclooctyne (DBCO) groups. It is designed for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) to enable bioconjugation without copper catalysts. The short PEG2 spacer increases solubility and flexibility, and the product is supplied with solubility and storage guidance for research use.

  • Supports copper-free click chemistry (SPAAC)
  • Contains two terminal DBCO reactive groups
  • PEG2 spacer improves aqueous solubility and flexibility
  • High reported purity suitable for research applications
  • Soluble in DMSO; in vitro and in vivo formulation protocols provided
  • Recommended storage as powder at -20°C for long-term stability

Catalog No. 50-002-86577 Supplier Medchemexpress LLC Supplier No. HY15183410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG2-DBCO is a bifunctional polyethylene glycol (PEG) linker bearing two terminal dibenzocyclooctyne (DBCO) groups. It is designed for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) to enable bioconjugation without copper catalysts. The short PEG2 spacer increases solubility and flexibility, and the product is supplied with solubility and storage guidance for research use.

  • Supports copper-free click chemistry (SPAAC)
  • Contains two terminal DBCO reactive groups
  • PEG2 spacer improves aqueous solubility and flexibility
  • High reported purity suitable for research applications
  • Soluble in DMSO; in vitro and in vivo formulation protocols provided
  • Recommended storage as powder at -20°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.