Learn More
Medchemexpress LLC DBCO-PEG2-NHS ester | 2585653-12-7 | 849001 | C30H31N3O8 | 25 MG
SDP
Catalog No. 5000374252
Encompass_Preferred
DBCO-PEG2-NHS ester is a click chemistry reagent that contains an azide group and an NHS ester. The NHS ester reacts specifically and efficiently with primary amines under neutral or slightly basic conditions to form a covalent bond. The hydrophilic PEG spacer arm enhances water solubility and provides a long, flexible connection to minimize steric hindrance. DBCO is used for copper-free Click Chemistry reactions. It is intended for research use only.
  • Contains an azide group and an NHS ester
  • NHS ester reacts specifically and efficiently with primary amines
  • Hydrophilic PEG spacer arm enhances water solubility
  • Minimizes steric hindrance in ligation
  • Used for copper-free click chemistry reactions
  • Reagent grade
  • For research use only
Catalog No. 50-003-74252 Supplier Medchemexpress LLC Supplier No. 25MGHY151827
DBCO-PEG2-NHS ester is a click chemistry reagent that contains an azide group and an NHS ester. The NHS ester reacts specifically and efficiently with primary amines under neutral or slightly basic conditions to form a covalent bond. The hydrophilic PEG spacer arm enhances water solubility and provides a long, flexible connection to minimize steric hindrance. DBCO is used for copper-free Click Chemistry reactions. It is intended for research use only.
  • Contains an azide group and an NHS ester
  • NHS ester reacts specifically and efficiently with primary amines
  • Hydrophilic PEG spacer arm enhances water solubility
  • Minimizes steric hindrance in ligation
  • Used for copper-free click chemistry reactions
  • Reagent grade
  • For research use only