Learn More
Medchemexpress LLC Dbco-peg2-val-cit-pab-mmae | 2845934-59-8 | 99.3% | 1569.92 g/mol | C84H120N12O17 | 25 MG
SDP
Catalog No. 5000202869
Encompass_Preferred
A cleavable ADC linker composed of a DBCO reactive handle, a PEG2 spacer, a cathepsin-cleavable valine-citrulline dipeptide with a para-aminobenzyl (PAB) self-immolative spacer, and the cytotoxic MMAE payload. Designed for conjugation via strain-promoted alkyne-azide cycloaddition (SPAAC) to form antibody-drug conjugates.
  • High purity suitable for conjugation reactions.
  • Cleavable val-cit dipeptide enables payload release in lysosomes.
  • Dbco moiety supports copper-free click conjugation (SPAAC).
  • Peg2 spacer improves solubility and linker flexibility.
  • Available in small mg-scale quantities for synthesis and development.
Catalog No. 50-002-02869 Supplier Medchemexpress LLC Supplier No. HY17228725MG
A cleavable ADC linker composed of a DBCO reactive handle, a PEG2 spacer, a cathepsin-cleavable valine-citrulline dipeptide with a para-aminobenzyl (PAB) self-immolative spacer, and the cytotoxic MMAE payload. Designed for conjugation via strain-promoted alkyne-azide cycloaddition (SPAAC) to form antibody-drug conjugates.
  • High purity suitable for conjugation reactions.
  • Cleavable val-cit dipeptide enables payload release in lysosomes.
  • Dbco moiety supports copper-free click conjugation (SPAAC).
  • Peg2 spacer improves solubility and linker flexibility.
  • Available in small mg-scale quantities for synthesis and development.