Learn More
Medchemexpress LLC Dbco-(PEG2-VC-PAB-MMAE)2 | 2259318-55-1 | 99.2% | 2,835.50 g/mol | C149H224N22O32 | 50 MG
SDP
Catalog No. 5000204131
Encompass_Preferred
DBCO-(PEG2-VC-PAB-MMAE)2 is a drug-linker conjugate for antibody-drug conjugate (ADC) research that combines a DBCO reactive group with a cleavable valine-citrulline linker, a p-aminobenzyl self-immolative spacer, and dual MMAE payloads. It is supplied as a purified powder for use in bio-conjugation via strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Provides a DBCO reactive group for copper-free click chemistry (SPAAC).
  • Includes a PEG2 spacer for improved solubility and flexibility.
  • Features a valine-citrulline cleavable linker with a p-aminobenzyl self-immolative spacer.
  • Contains two MMAE payloads for potent cytotoxic delivery.
  • High purity, typically 99.2%.
  • Supplied as a powder suitable for ADC construction and conjugation.
Catalog No. 50-002-04131 Supplier Medchemexpress LLC Supplier No. HY12669050MG
DBCO-(PEG2-VC-PAB-MMAE)2 is a drug-linker conjugate for antibody-drug conjugate (ADC) research that combines a DBCO reactive group with a cleavable valine-citrulline linker, a p-aminobenzyl self-immolative spacer, and dual MMAE payloads. It is supplied as a purified powder for use in bio-conjugation via strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Provides a DBCO reactive group for copper-free click chemistry (SPAAC).
  • Includes a PEG2 spacer for improved solubility and flexibility.
  • Features a valine-citrulline cleavable linker with a p-aminobenzyl self-immolative spacer.
  • Contains two MMAE payloads for potent cytotoxic delivery.
  • High purity, typically 99.2%.
  • Supplied as a powder suitable for ADC construction and conjugation.