Learn More
Medchemexpress LLC DBCO-PEG24-maleimide | 2924872-84-2 | 98.0% | 1555.81 | C76H122N4O29 | 5mg
SDP
Catalog No. 5000270290
Encompass_Preferred
DBCO-PEG24-Maleimide contains a maleimide and a DBCO group DBCO-PEG24-Maleimide can be used for thiol-containing biomolecule conjugations DBCO-PEG24-Maleimide can be used in click chemistry with azide-bearing biomolecules[1]
Catalog No. 50-002-70290 Supplier Medchemexpress LLC Supplier No. HYW8790485MG
DBCO-PEG24-Maleimide contains a maleimide and a DBCO group DBCO-PEG24-Maleimide can be used for thiol-containing biomolecule conjugations DBCO-PEG24-Maleimide can be used in click chemistry with azide-bearing biomolecules[1]