Learn More
Medchemexpress LLC DBCO-PEG24-NHS ester | 2765066-38-2 | MFCD34551889 | 95.5% | 1530.74 | C74H119N3O30 | 10 MG
SDP
Catalog No. 5000285888
Encompass_Preferred
DBCO-PEG24-NHS ester is a bifunctional polyethylene glycol (PEG) reagent containing a dibenzocyclooctyne (DBCO) group and an N-hydroxysuccinimide (NHS) ester. It is designed for bioconjugation workflows where copper-free click chemistry and selective amine coupling are required; the PEG24 spacer increases water solubility and reduces steric hindrance. Supplied for research use with documented purity and recommended storage conditions.
  • Enables copper-free click chemistry via DBCO-mediated SPAAC.
  • Reacts selectively with primary amines through NHS ester chemistry.
  • Provides a PEG24 spacer to increase water solubility and flexibility.
  • Supplied with documented purity and certificate of analysis.
  • Stable when stored sealed, away from moisture and light under recommended temperatures.
  • Available in small research-scale quantities.
Catalog No. 50-002-85888 Supplier Medchemexpress LLC Supplier No. HY15183510MG
DBCO-PEG24-NHS ester is a bifunctional polyethylene glycol (PEG) reagent containing a dibenzocyclooctyne (DBCO) group and an N-hydroxysuccinimide (NHS) ester. It is designed for bioconjugation workflows where copper-free click chemistry and selective amine coupling are required; the PEG24 spacer increases water solubility and reduces steric hindrance. Supplied for research use with documented purity and recommended storage conditions.
  • Enables copper-free click chemistry via DBCO-mediated SPAAC.
  • Reacts selectively with primary amines through NHS ester chemistry.
  • Provides a PEG24 spacer to increase water solubility and flexibility.
  • Supplied with documented purity and certificate of analysis.
  • Stable when stored sealed, away from moisture and light under recommended temperatures.
  • Available in small research-scale quantities.