Learn More
Medchemexpress LLC Dbco-peg3-acid | 2754384-59-1 | 98.35% | 508.56 | 5 MG
SDP
Catalog No. 5000403589
Encompass_Preferred
DBCO-PEG3-acid is a non-cleavable 3 unit PEG ADC linker used in the synthesis of antibody-drug conjugates. It contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • Non-cleavable linker
Catalog No. 50-004-03589 Supplier Medchemexpress LLC Supplier No. HY429745MG
DBCO-PEG3-acid is a non-cleavable 3 unit PEG ADC linker used in the synthesis of antibody-drug conjugates. It contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • Non-cleavable linker