Learn More
Medchemexpress LLC Dbco-peg3-acid | 2754384-59-1 | 98.4% | 508.56 g/mol | C28H32N2O7 | 10 MG
SDP
Catalog No. 5000299082
Encompass_Preferred
DBCO-PEG3-acid is a dibenzocyclooctyne (DBCO)-containing PEG linker designed for copper-free click chemistry (strain-promoted alkyne-azide cycloaddition) in bioconjugation and antibody-drug conjugate synthesis. It is a non-cleavable linker supplied as a solid for laboratory use.
Catalog No. 50-002-99082 Supplier Medchemexpress LLC Supplier No. HY4297410MG
DBCO-PEG3-acid is a dibenzocyclooctyne (DBCO)-containing PEG linker designed for copper-free click chemistry (strain-promoted alkyne-azide cycloaddition) in bioconjugation and antibody-drug conjugate synthesis. It is a non-cleavable linker supplied as a solid for laboratory use.