Learn More
Medchemexpress LLC DBCO-PEG3-Glu-VC-PABC-MMAF | 2253947-24-7 | 1 MG
SDP
Catalog No. 5000339850
Encompass_Preferred
DBCO-PEG3-Glu-VC-PABC-MMAF is a drug-linker conjugate for Antibody-Drug Conjugates (ADCs). It utilizes the tubulin inhibitor MMAF, linked via a cathepsin cleavable DBCO-PEG3-Glu-VC-PABC. This compound can be used in the synthesis of ADCs, which consist of an antibody to which an ADC cytotoxin is attached through an ADC linker.
  • Drug-linker conjugate for Antibody-Drug Conjugates (ADCs)
  • Uses tubulin inhibitor MMAF
  • Linked via cathepsin cleavable DBCO-PEG3-Glu-VC-PABC
  • Can be used in the synthesis of ADCs
  • For research use only
Catalog No. 50-003-39850 Supplier Medchemexpress LLC Supplier No. HY1311581MG
DBCO-PEG3-Glu-VC-PABC-MMAF is a drug-linker conjugate for Antibody-Drug Conjugates (ADCs). It utilizes the tubulin inhibitor MMAF, linked via a cathepsin cleavable DBCO-PEG3-Glu-VC-PABC. This compound can be used in the synthesis of ADCs, which consist of an antibody to which an ADC cytotoxin is attached through an ADC linker.
  • Drug-linker conjugate for Antibody-Drug Conjugates (ADCs)
  • Uses tubulin inhibitor MMAF
  • Linked via cathepsin cleavable DBCO-PEG3-Glu-VC-PABC
  • Can be used in the synthesis of ADCs
  • For research use only