Learn More
Medchemexpress LLC Dbco-peg3-vc-exatecan | 2754384-70-6 | 96.7% | 1331.44 | C71H79FN10O15 | 5 MG
SDP
Catalog No. 5000202482
Encompass_Preferred
DBCO-PEG3-VC-Exatecan is a research-grade drug-linker conjugate that couples a cleavable DBCO-PEG3-VC linker to the topoisomerase I inhibitor exatecan, supplied as an off-white to light yellow solid for use in antibody-drug conjugate (ADC) research.
  • adc drug-linker conjugate for preclinical and conjugation work
  • contains exatecan, a DNA topoisomerase I inhibitor
  • cleavable DBCO-PEG3-VC linker suitable for bioconjugation
  • high reported purity for research applications
  • available in small milligram quantities for formulation and testing
Catalog No. 50-002-02482 Supplier Medchemexpress LLC Supplier No. HY1647925MG
DBCO-PEG3-VC-Exatecan is a research-grade drug-linker conjugate that couples a cleavable DBCO-PEG3-VC linker to the topoisomerase I inhibitor exatecan, supplied as an off-white to light yellow solid for use in antibody-drug conjugate (ADC) research.
  • adc drug-linker conjugate for preclinical and conjugation work
  • contains exatecan, a DNA topoisomerase I inhibitor
  • cleavable DBCO-PEG3-VC linker suitable for bioconjugation
  • high reported purity for research applications
  • available in small milligram quantities for formulation and testing