Learn More
MOLECULAR DEPOT LLC DBCO-PEG36 Beads - 1 mL
Catalog No. 5000013375
Encompass_Preferred
DBCO-PEG36 Magnetic Beads (1 um) is a suspension of magnetic beads ( 1 um in diameter 0 5perc 5 mg/mL) that are functionalized with a DBCO (Dibenzocyclooctyne) group and a polyethylene glycol (PEG) spacer of 36 units in length This product can covalently capture azide-tagged molecules using the so-called Click Chemistry For in vitro research use only
Catalog No. 50-000-13375 Supplier MOLECULAR DEPOT LLC Supplier No. B2021917
DBCO-PEG36 Magnetic Beads (1 um) is a suspension of magnetic beads ( 1 um in diameter 0 5perc 5 mg/mL) that are functionalized with a DBCO (Dibenzocyclooctyne) group and a polyethylene glycol (PEG) spacer of 36 units in length This product can covalently capture azide-tagged molecules using the so-called Click Chemistry For in vitro research use only