Learn More
Medchemexpress LLC Dbco-peg4-acetic-val-cit-pab | 98.2% | 855.97 | C45H57N7O10 | 50 MG
SDP
Catalog No. 5000202550
Encompass_Preferred
DBCO-PEG4-acetic-Val-Cit-PAB is a cleavable four-unit polyethylene glycol (PEG) linker that contains a dibenzocyclooctyne (DBCO) moiety and a Val-Cit-PAB protease-cleavable spacer. It is designed for use in antibody-drug conjugate assembly and for copper-free click chemistry (SPAAC) to attach azide-bearing payloads.
  • Cleavable Val-Cit-PAB dipeptide spacer for protease-triggered release.
  • Dbco group enables copper-free click conjugation (SPAAC) with azides.
  • Four-unit PEG spacer improves solubility and reduces steric hindrance.
  • High reported purity and defined molecular weight support reproducible conjugation chemistry.
Catalog No. 50-002-02550 Supplier Medchemexpress LLC Supplier No. HY13609850MG
DBCO-PEG4-acetic-Val-Cit-PAB is a cleavable four-unit polyethylene glycol (PEG) linker that contains a dibenzocyclooctyne (DBCO) moiety and a Val-Cit-PAB protease-cleavable spacer. It is designed for use in antibody-drug conjugate assembly and for copper-free click chemistry (SPAAC) to attach azide-bearing payloads.
  • Cleavable Val-Cit-PAB dipeptide spacer for protease-triggered release.
  • Dbco group enables copper-free click conjugation (SPAAC) with azides.
  • Four-unit PEG spacer improves solubility and reduces steric hindrance.
  • High reported purity and defined molecular weight support reproducible conjugation chemistry.