Learn More
Medchemexpress LLC DBCO-PEG4-Biotin | 1255942-07-4 | >99.4% | 1 ML
SDP
Catalog No. 5000340658
Encompass_Preferred
DBCO-PEG4-Biotin is an azadibenzocyclooctyne-biotin derivative that includes a biotin group and four PEG units. It functions as a versatile biotinylation reagent, enabling the introduction of a biotin moiety to azide-labeled biomolecules through copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reactions. The DBCO group within DBCO-PEG4-Biotin facilitates SPAAC with molecules containing azide groups.
  • Versatile biotinylation reagent
  • Introduces biotin moiety to azide-labeled biomolecules
  • Utilizes copper-free strain-promoted alkyne-azide click chemistry (SPAAC)
  • DBCO group facilitates SPAAC with azide groups
Catalog No. 50-003-40658 Supplier Medchemexpress LLC Supplier No. HY13080910MM/1ML
DBCO-PEG4-Biotin is an azadibenzocyclooctyne-biotin derivative that includes a biotin group and four PEG units. It functions as a versatile biotinylation reagent, enabling the introduction of a biotin moiety to azide-labeled biomolecules through copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reactions. The DBCO group within DBCO-PEG4-Biotin facilitates SPAAC with molecules containing azide groups.
  • Versatile biotinylation reagent
  • Introduces biotin moiety to azide-labeled biomolecules
  • Utilizes copper-free strain-promoted alkyne-azide click chemistry (SPAAC)
  • DBCO group facilitates SPAAC with azide groups