Learn More
Medchemexpress LLC DBCO-PEG4-Biotin | 1255942-07-4 | 99.5% | 100 MG
SDP
Catalog No. 5000339000
Encompass_Preferred
DBCO-PEG4-Biotin is an azadibenzocyclooctyne-biotin derivative that features a biotin group and four polyethylene glycol (PEG) units. This versatile biotinylation reagent is utilized for the introduction of a biotin moiety to azide-labeled biomolecules. It enables copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reactions, leading to the formation of a stable triazole linkage.
  • Contains a biotin group and four PEG units.
  • Introduces a biotin moiety to azide-labeled biomolecules.
  • Facilitates copper-free strain-promoted alkyne-azide click chemistry.
  • Forms a stable triazole linkage.
  • Utilized for research purposes.
Catalog No. 50-003-39000 Supplier Medchemexpress LLC Supplier No. HY130809100MG
DBCO-PEG4-Biotin is an azadibenzocyclooctyne-biotin derivative that features a biotin group and four polyethylene glycol (PEG) units. This versatile biotinylation reagent is utilized for the introduction of a biotin moiety to azide-labeled biomolecules. It enables copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reactions, leading to the formation of a stable triazole linkage.
  • Contains a biotin group and four PEG units.
  • Introduces a biotin moiety to azide-labeled biomolecules.
  • Facilitates copper-free strain-promoted alkyne-azide click chemistry.
  • Forms a stable triazole linkage.
  • Utilized for research purposes.