Learn More
Medchemexpress LLC DBCO-PEG4-Biotin | 1255942-07-4 | 99.5% | 25 MG
SDP
Catalog No. 5000342771
Encompass_Preferred
DBCO-PEG4-Biotin is an azadibenzocyclooctyne-biotin derivative containing a biotin group and 4 PEGs. It is a versatile biotinylation reagent used for introducing a biotin moiety to azide-labeled biomolecules via copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reaction. This reagent contains a DBCO group that can undergo SPAAC with molecules containing azide groups.
  • Azadibenzocyclooctyne-biotin derivative
  • Contains a biotin group and 4 PEGs
  • Versatile biotinylation reagent
  • Introduces a biotin moiety to azide-labeled biomolecules
  • Uses copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reaction
  • DBCO group can undergo SPAAC with molecules containing azide groups
Catalog No. 50-003-42771 Supplier Medchemexpress LLC Supplier No. 25MGHY130809
DBCO-PEG4-Biotin is an azadibenzocyclooctyne-biotin derivative containing a biotin group and 4 PEGs. It is a versatile biotinylation reagent used for introducing a biotin moiety to azide-labeled biomolecules via copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reaction. This reagent contains a DBCO group that can undergo SPAAC with molecules containing azide groups.
  • Azadibenzocyclooctyne-biotin derivative
  • Contains a biotin group and 4 PEGs
  • Versatile biotinylation reagent
  • Introduces a biotin moiety to azide-labeled biomolecules
  • Uses copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reaction
  • DBCO group can undergo SPAAC with molecules containing azide groups