Learn More
Medchemexpress LLC DBCO-PEG4-Biotin | 1255942-07-4 | MFCD22572661 | 99.4% | C39H51N5O8S | 10MG
SDP
Catalog No. 5000261811
Encompass_Preferred
DBCO-PEG4-Biotin is a dibenzocyclooctyne-linked biotin reagent with a PEG4 spacer designed for copper-free click biotinylation of azide-labeled biomolecules. It enables rapid strain-promoted azide-alkyne cycloaddition to form a stable triazole linkage and introduces a biotin handle for affinity capture or detection in labeling and pull-down applications.
  • Enables copper-free strain-promoted azide-alkyne cycloaddition (SPAAC) with azide-labeled substrates.
  • Introduces a biotin affinity tag for streptavidin-based capture or detection.
  • Peg4 spacer improves solubility and reduces steric hindrance during conjugation.
  • High purity suitable for sensitive bioconjugation workflows.
  • Offered as solid and as a DMSO solution for flexible experimental use.
  • Stable when stored sealed and away from moisture under recommended conditions.
Catalog No. 50-002-61811 Supplier Medchemexpress LLC Supplier No. HY13080910MG
DBCO-PEG4-Biotin is a dibenzocyclooctyne-linked biotin reagent with a PEG4 spacer designed for copper-free click biotinylation of azide-labeled biomolecules. It enables rapid strain-promoted azide-alkyne cycloaddition to form a stable triazole linkage and introduces a biotin handle for affinity capture or detection in labeling and pull-down applications.
  • Enables copper-free strain-promoted azide-alkyne cycloaddition (SPAAC) with azide-labeled substrates.
  • Introduces a biotin affinity tag for streptavidin-based capture or detection.
  • Peg4 spacer improves solubility and reduces steric hindrance during conjugation.
  • High purity suitable for sensitive bioconjugation workflows.
  • Offered as solid and as a DMSO solution for flexible experimental use.
  • Stable when stored sealed and away from moisture under recommended conditions.