Learn More
Medchemexpress LLC HY-130809 5mg Medchemexpress, DBCO-PEG4-Biotin CAS:1255942-07-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1308095MG
Encompass_Preferred
Medchemexpress, HY-130809 5mg DBCO-PEG4-Biotin CAS:1255942-07-4 DBCO-PEG4-Biotin is an azadibenzocyclooctyne-biotin derivative containing a biotin group and 4 PEGs. DBCO-PEG4-Biotin is a versatile biotinylation reagent used for the introduction of a biotin moiety to azide-labeled biomolecules via copper-free strain-promoted alkyne-azide click chemistry (SPAAC) reaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1253