Learn More
Medchemexpress LLC DBCO-PEG4-DBCO | 2182601-68-7 | 97.4% | 1 G
SDP
Catalog No. 5000340918
Encompass_Preferred
DBCO-PEG4-DBCO is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a cleavable ADC linker for synthesizing antibody-drug conjugates (ADCs) and is a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • PEG-based PROTAC linker
  • Cleavable ADC linker
  • Click chemistry reagent
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-40918 Supplier Medchemexpress LLC Supplier No. HY1303461G
DBCO-PEG4-DBCO is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a cleavable ADC linker for synthesizing antibody-drug conjugates (ADCs) and is a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • PEG-based PROTAC linker
  • Cleavable ADC linker
  • Click chemistry reagent
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)