Learn More
Medchemexpress LLC DBCO-PEG4-DBCO | 2182601-68-7 | 97.4% | 250 MG
SDP
Catalog No. 5000342492
Encompass_Preferred
DBCO-PEG4-DBCO is a versatile PEG-based PROTAC linker and a cleavable ADC linker. It is utilized in the synthesis of PROTACs and antibody-drug conjugates (ADCs). This compound functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Used in PROTAC synthesis
  • Cleavable ADC linker
  • Click chemistry reagent
  • Features a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide groups
Catalog No. 50-003-42492 Supplier Medchemexpress LLC Supplier No. 250MGHY130346
DBCO-PEG4-DBCO is a versatile PEG-based PROTAC linker and a cleavable ADC linker. It is utilized in the synthesis of PROTACs and antibody-drug conjugates (ADCs). This compound functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Used in PROTAC synthesis
  • Cleavable ADC linker
  • Click chemistry reagent
  • Features a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide groups