Learn More
Medchemexpress LLC DBCO-PEG4-DBCO | 2182601-68-7 | 97.4% | 50 MG
SDP
Catalog No. 5000340917
Encompass_Preferred
DBCO-PEG4-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a cleavable ADC linker for the creation of antibody-drug conjugates (ADCs). This compound is a click chemistry reagent, featuring a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Cleavable ADC linker.
  • Used in the synthesis of antibody-drug conjugates (ADCs).
  • Click chemistry reagent containing a DBCO group.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
Catalog No. 50-003-40917 Supplier Medchemexpress LLC Supplier No. HY13034650MG
DBCO-PEG4-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a cleavable ADC linker for the creation of antibody-drug conjugates (ADCs). This compound is a click chemistry reagent, featuring a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Cleavable ADC linker.
  • Used in the synthesis of antibody-drug conjugates (ADCs).
  • Click chemistry reagent containing a DBCO group.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.