Learn More
Medchemexpress LLC DBCO-PEG4-DBCO | 2182601-68-7 | 97.4% | 810.93 | C48H50N4O8 | 10 MG
SDP
Catalog No. 5000203235
Encompass_Preferred
DBCO-PEG4-DBCO is a PEG-based bifunctional linker bearing two dibenzocyclooctyne (DBCO) groups. It is used as a click chemistry reagent for strain-promoted alkyne-azide cycloaddition (SPAAC) and as a PEG linker in PROTAC and antibody-drug conjugate (ADC) synthesis.
  • Click chemistry reagent enabling SPAAC with azide-functionalized partners.
  • Provides a PEG4 spacer to improve solubility and linker flexibility.
  • High solubility in DMSO; manufacturer provides in vitro and in vivo formulation protocols.
  • Typical purity around 97.4% and appears as a white to yellow solid.
  • Suitable for use in PROTAC and ADC synthesis workflows.
Catalog No. 50-002-03235 Supplier Medchemexpress LLC Supplier No. HY13034610MG
DBCO-PEG4-DBCO is a PEG-based bifunctional linker bearing two dibenzocyclooctyne (DBCO) groups. It is used as a click chemistry reagent for strain-promoted alkyne-azide cycloaddition (SPAAC) and as a PEG linker in PROTAC and antibody-drug conjugate (ADC) synthesis.
  • Click chemistry reagent enabling SPAAC with azide-functionalized partners.
  • Provides a PEG4 spacer to improve solubility and linker flexibility.
  • High solubility in DMSO; manufacturer provides in vitro and in vivo formulation protocols.
  • Typical purity around 97.4% and appears as a white to yellow solid.
  • Suitable for use in PROTAC and ADC synthesis workflows.