Learn More
Medchemexpress LLC DBCO-PEG4-Desthiobiotin | 2032788-37-5 | 96.4% | C39H53N5O8 | 25 MG
SDP
Catalog No. 5000352571
Encompass_Preferred
DBCO-PEG4-Desthiobiotin is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring a DBCO group that enables strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide groups
  • For research use only
Catalog No. 50-003-52571 Supplier Medchemexpress LLC Supplier No. 25MGHY140301
DBCO-PEG4-Desthiobiotin is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring a DBCO group that enables strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide groups
  • For research use only