Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-PEG4-GGFG-Dxd | 2694856-51-2 | 98.4% | 1375.45 | C72H79FN10O17 | 100 MG
SDP

Catalog No. 5000203446
Encompass_Preferred

DBCO-PEG4-GGFG-Dxd is a DBCO-terminated PEG4-linked GGFG drug-linker conjugate that carries Dxd, a DNA topoisomerase I inhibitor. It is supplied as a solid for research use and is intended for antibody-drug conjugate (ADC) development and click-chemistry conjugation via strain-promoted alkyne-azide cycloaddition (SPAAC).

  • Chemical formula: C72H79FN10O17.
  • Molecular weight: 1375.45 g/mol.
  • Purity: 98.4%.
  • Appearance: off-white to light yellow solid.
  • CAS number: 2694856-51-2.
  • Storage: -20°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Solubility: DMSO 50 mg/mL (36.35 mM) with warming/sonication.
  • Intended use: for research use only; not for clinical applications.

Catalog No. 50-002-03446 Supplier Medchemexpress LLC Supplier No. HY134723100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG4-GGFG-Dxd is a DBCO-terminated PEG4-linked GGFG drug-linker conjugate that carries Dxd, a DNA topoisomerase I inhibitor. It is supplied as a solid for research use and is intended for antibody-drug conjugate (ADC) development and click-chemistry conjugation via strain-promoted alkyne-azide cycloaddition (SPAAC).

  • Chemical formula: C72H79FN10O17.
  • Molecular weight: 1375.45 g/mol.
  • Purity: 98.4%.
  • Appearance: off-white to light yellow solid.
  • CAS number: 2694856-51-2.
  • Storage: -20°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Solubility: DMSO 50 mg/mL (36.35 mM) with warming/sonication.
  • Intended use: for research use only; not for clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.