Learn More
Medchemexpress LLC Dbco-peg4-ggfg-dxd | 2694856-51-2 | MFCD22575025 | 98.4% | 1375.47 | C72H79FN10O17 | 5 MG
SDP
Catalog No. 5000202036
Encompass_Preferred
DBCO-PEG4-GGFG-Dxd is an agent-linker conjugate used in antibody-drug conjugate (ADC) research that incorporates a cleavable GGFG linker and a DBCO-PEG4 moiety to enable strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry for conjugation to azide-containing molecules.
  • Provides a payload-linker suitable for ADC construction.
  • Enables copper-free SPAAC click chemistry with azide-functionalized partners.
  • Contains a cleavable GGFG linker for payload release.
  • Supplied as an off-white to light yellow solid for ease of handling.
  • Stable when stored protected from light at -20°C; in solvent stability is extended at -80°C.
Catalog No. 50-002-02036 Supplier Medchemexpress LLC Supplier No. HY1347235MG
DBCO-PEG4-GGFG-Dxd is an agent-linker conjugate used in antibody-drug conjugate (ADC) research that incorporates a cleavable GGFG linker and a DBCO-PEG4 moiety to enable strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry for conjugation to azide-containing molecules.
  • Provides a payload-linker suitable for ADC construction.
  • Enables copper-free SPAAC click chemistry with azide-functionalized partners.
  • Contains a cleavable GGFG linker for payload release.
  • Supplied as an off-white to light yellow solid for ease of handling.
  • Stable when stored protected from light at -20°C; in solvent stability is extended at -80°C.