Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbco-PEG4-GGFG-Dxd | 98.4% | 1375.45 g/mol | C72H79FN10O17 | 50 MG
SDP

Catalog No. 5000203338
Encompass_Preferred

DBCO-PEG4-GGFG-Dxd is a DBCO-functionalized PEG4-linked GGFG peptide drug-linker conjugate incorporating Dxd, a topoisomerase I inhibitor, designed for antibody-drug conjugate (ADC) research and click-chemistry conjugation via its DBCO handle.

  • Designed for ADC research and click-chemistry conjugation
  • Contains Dxd, a topoisomerase I inhibitor
  • Purity 98.4%
  • Molecular weight 1375.45 g/mol
  • Chemical formula C72H79FN10O17
  • Appearance: solid, off-white to light yellow
  • Storage: store protected from light; in solvent -80°C (6 months), -20°C (1 month)
  • Available in small research quantities including 50 MG

Catalog No. 50-002-03338 Supplier Medchemexpress LLC Supplier No. HY13472350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG4-GGFG-Dxd is a DBCO-functionalized PEG4-linked GGFG peptide drug-linker conjugate incorporating Dxd, a topoisomerase I inhibitor, designed for antibody-drug conjugate (ADC) research and click-chemistry conjugation via its DBCO handle.

  • Designed for ADC research and click-chemistry conjugation
  • Contains Dxd, a topoisomerase I inhibitor
  • Purity 98.4%
  • Molecular weight 1375.45 g/mol
  • Chemical formula C72H79FN10O17
  • Appearance: solid, off-white to light yellow
  • Storage: store protected from light; in solvent -80°C (6 months), -20°C (1 month)
  • Available in small research quantities including 50 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.