Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-PEG4-maleimide | 1480516-75-3 | 95.9% | 674.74 g/mol | C36H42N4O9 | 10 MG
SDP

Catalog No. 5000204276
Encompass_Preferred

DBCO-PEG4-maleimide is a bifunctional PEG4 linker bearing a strained dibenzocyclooctyne (DBCO) and a terminal maleimide. It enables site-selective conjugation by combining thiol-maleimide chemistry with strain-promoted azide-alkyne cycloaddition, making it suitable for antibody-drug conjugate assembly and other bioconjugation workflows.

  • Enables site-selective conjugation via thiol-maleimide coupling and SPAAC
  • Contains a PEG4 spacer for increased solubility and reduced steric hindrance
  • Liquid form, colorless to light yellow, suitable for solution-phase reactions
  • High reported purity supports reproducible conjugation results
  • Soluble in DMSO at approximately 50 mg/mL; ultrasonic assistance may be required
  • Store protected from light; keep frozen under inert atmosphere for long-term storage

Catalog No. 50-002-04276 Supplier Medchemexpress LLC Supplier No. HY12077010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG4-maleimide is a bifunctional PEG4 linker bearing a strained dibenzocyclooctyne (DBCO) and a terminal maleimide. It enables site-selective conjugation by combining thiol-maleimide chemistry with strain-promoted azide-alkyne cycloaddition, making it suitable for antibody-drug conjugate assembly and other bioconjugation workflows.

  • Enables site-selective conjugation via thiol-maleimide coupling and SPAAC
  • Contains a PEG4 spacer for increased solubility and reduced steric hindrance
  • Liquid form, colorless to light yellow, suitable for solution-phase reactions
  • High reported purity supports reproducible conjugation results
  • Soluble in DMSO at approximately 50 mg/mL; ultrasonic assistance may be required
  • Store protected from light; keep frozen under inert atmosphere for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.