Learn More
Medchemexpress LLC Dbco-peg4-nhs ester | 1427004-19-0 | 99.0% | C34H39N3O10 | 250 MG
SDP
Catalog No. 5000352875
Encompass_Preferred
DBCO-PEG4-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, incorporating a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Peg-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • PROTACs are composed of two ligands linked together: one for an E3 ubiquitin ligase and another for a target protein.
  • PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
Catalog No. 50-003-52875 Supplier Medchemexpress LLC Supplier No. 250MGHY140272
DBCO-PEG4-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, incorporating a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Peg-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • PROTACs are composed of two ligands linked together: one for an E3 ubiquitin ligase and another for a target protein.
  • PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.