Learn More
TARGETMOL CHEMICALS INC DBCO-PEG4-NHS ESTER 5MG
SDP
Catalog No. 502581255
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DBCO-PEG4-NHS ester is a PEG-based PROTAC linker that can be used in PROTAC synthesis[1]. purity: 95%
Catalog No. 50-258-1255 Supplier TARGETMOL CHEMICALS INC Supplier No. T150735MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DBCO-PEG4-NHS ester is a PEG-based PROTAC linker that can be used in PROTAC synthesis[1]. purity: 95%