Learn More
Medchemexpress LLC DBCO-PEG4-PFP ester | 00-00-0 | 95.0% | C36H35F5N2O8 | 10MG
SDP
Catalog No. 5000261451
Encompass_Preferred
DBCO-PEG4-PFP ester is a PEG-based linker and click-chemistry reagent used for bioconjugation and PROTAC linker construction. It combines a dibenzocyclooctyne (DBCO) group for strain-promoted alkyne-azide cycloaddition (SPAAC) with a pentafluorophenyl (PFP) ester reactive handle for coupling to nucleophiles such as amines.
  • Enables copper-free click reactions with azide-functionalized partners.
  • Provides an active pentafluorophenyl ester for efficient amide bond formation with amines.
  • Includes a PEG4 spacer to improve solubility and reduce steric hindrance.
  • Suitable for PROTAC linker synthesis and general bioconjugation workflows.
  • High purity and recommended storage support reproducible laboratory results.
Catalog No. 50-002-61451 Supplier Medchemexpress LLC Supplier No. HY14027810MG
DBCO-PEG4-PFP ester is a PEG-based linker and click-chemistry reagent used for bioconjugation and PROTAC linker construction. It combines a dibenzocyclooctyne (DBCO) group for strain-promoted alkyne-azide cycloaddition (SPAAC) with a pentafluorophenyl (PFP) ester reactive handle for coupling to nucleophiles such as amines.
  • Enables copper-free click reactions with azide-functionalized partners.
  • Provides an active pentafluorophenyl ester for efficient amide bond formation with amines.
  • Includes a PEG4 spacer to improve solubility and reduce steric hindrance.
  • Suitable for PROTAC linker synthesis and general bioconjugation workflows.
  • High purity and recommended storage support reproducible laboratory results.