Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbco-peg4-pfp ester | 2182601-19-8 | 95.0% | C36H35F5N2O8 | 100 MG
SDP

Catalog No. 5000353140
Encompass_Preferred

DBCO-PEG4-PFP ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. This allows it to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.

  • Peg-based protac linker
  • Utilized in protac synthesis
  • Functions as a click chemistry reagent
  • Contains a dbco group for spaac reactions
  • Facilitates selective degradation of target proteins
  • Appears as a colorless to light yellow oil
  • Purity of 95.0%
  • Molecular weight: 718.66
  • Molecular formula: C36H35F5N2O8

Catalog No. 50-003-53140 Supplier Medchemexpress LLC Supplier No. HY140278100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG4-PFP ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. This allows it to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.

  • Peg-based protac linker
  • Utilized in protac synthesis
  • Functions as a click chemistry reagent
  • Contains a dbco group for spaac reactions
  • Facilitates selective degradation of target proteins
  • Appears as a colorless to light yellow oil
  • Purity of 95.0%
  • Molecular weight: 718.66
  • Molecular formula: C36H35F5N2O8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.