Learn More
Medchemexpress LLC DBCO-PEG4-PFP ester | 2182601-19-8 | 95.0% | C36H35F5N2O8 | 50 MG
SDP
Catalog No. 5000357333
Encompass_Preferred
DBCO-PEG4-PFP ester is a PEG-based PROTAC linker that acts as a click chemistry reagent. It features a DBCO group designed for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules, making it suitable for PROTAC synthesis.
  • Utilized as a PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide-containing molecules
  • Supports synthesis of PROTACs for target protein degradation
Catalog No. 50-003-57333 Supplier Medchemexpress LLC Supplier No. HY14027850MG
DBCO-PEG4-PFP ester is a PEG-based PROTAC linker that acts as a click chemistry reagent. It features a DBCO group designed for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules, making it suitable for PROTAC synthesis.
  • Utilized as a PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide-containing molecules
  • Supports synthesis of PROTACs for target protein degradation