Learn More
Medchemexpress LLC DBCO-PEG4-PFP ester | 95.0% | C36H35F5N2O8 | 25 MG
SDP
Catalog No. 5000357388
Encompass_Preferred
DBCO-PEG4-PFP ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs are composed of two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other the protein of interest. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in protac synthesis
  • Functions as a click chemistry reagent
  • Incorporates a dbco group
  • Undergoes strain-promoted alkyne-azide cycloaddition (spaac) with azide groups
  • Leverages the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57388 Supplier Medchemexpress LLC Supplier No. 25MGHY140278
DBCO-PEG4-PFP ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs are composed of two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other the protein of interest. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in protac synthesis
  • Functions as a click chemistry reagent
  • Incorporates a dbco group
  • Undergoes strain-promoted alkyne-azide cycloaddition (spaac) with azide groups
  • Leverages the ubiquitin-proteasome system
  • Selectively degrades target proteins