Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbco-peg4-va-pbd | 2241644-09-5 | >98.0% | 1430.6 | C80H87N9O16 | 10 MG
SDP

Catalog No. 5000202559
Encompass_Preferred

DBCO-PEG4-VA-PBD is a drug-linker conjugate designed for antibody-drug conjugate (ADC) research and click chemistry applications. It combines a dibenzocyclooctyne (DBCO) group for strain-promoted alkyne-azide cycloaddition (SPAAC) with a PEG4-valine-alanine spacer and a pyrrolobenzodiazepine (PBD) cytotoxic payload. The compound is supplied for research use and is handled under low-temperature, inert storage conditions.

  • Contains DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Includes PEG4-valine-alanine spacer to provide linker stability.
  • Delivers a pyrrolobenzodiazepine (PBD) cytotoxic payload for ADC applications.
  • Molecular weight approximately 1,430.6 Da.
  • Soluble in DMSO at 120 mg/mL; ultrasonic agitation recommended.
  • Store at -80°C under nitrogen to maintain stability.

Catalog No. 50-002-02559 Supplier Medchemexpress LLC Supplier No. HY13343310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG4-VA-PBD is a drug-linker conjugate designed for antibody-drug conjugate (ADC) research and click chemistry applications. It combines a dibenzocyclooctyne (DBCO) group for strain-promoted alkyne-azide cycloaddition (SPAAC) with a PEG4-valine-alanine spacer and a pyrrolobenzodiazepine (PBD) cytotoxic payload. The compound is supplied for research use and is handled under low-temperature, inert storage conditions.

  • Contains DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Includes PEG4-valine-alanine spacer to provide linker stability.
  • Delivers a pyrrolobenzodiazepine (PBD) cytotoxic payload for ADC applications.
  • Molecular weight approximately 1,430.6 Da.
  • Soluble in DMSO at 120 mg/mL; ultrasonic agitation recommended.
  • Store at -80°C under nitrogen to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.