Learn More
Medchemexpress LLC Dbco-PEG4-Val-Cit-PAB-MMAF | 2244602-23-9 | 98.7% | 1672.01 | C88H126N12O20 | 10 MG
SDP
Catalog No. 5000203343
Encompass_Preferred
DBCO-PEG4-Val-Cit-PAB-MMAF is a cleavable ADC linker-payload conjugate that pairs a DBCO-functionalized PEG4 Val-Cit-PAB linker with the tubulin inhibitor MMAF. It is provided as a solid for use in antibody-drug conjugate synthesis and copper-free click chemistry (SPAAC) with azide-containing biomolecules. The Val-Cit-PAB spacer is designed for cathepsin-mediated cleavage to release the payload under lysosomal conditions.
  • Cleavable Val-Cit-PAB spacer for enzymatic release.
  • DBCO handle for copper-free click (SPAAC) conjugation.
  • Hydrophilic PEG4 spacer improves solubility and flexibility.
  • High reported purity suitable for research conjugation workflows.
  • Available in small milligram pack sizes for lab-scale synthesis.
Catalog No. 50-002-03343 Supplier Medchemexpress LLC Supplier No. HY13099010MG
DBCO-PEG4-Val-Cit-PAB-MMAF is a cleavable ADC linker-payload conjugate that pairs a DBCO-functionalized PEG4 Val-Cit-PAB linker with the tubulin inhibitor MMAF. It is provided as a solid for use in antibody-drug conjugate synthesis and copper-free click chemistry (SPAAC) with azide-containing biomolecules. The Val-Cit-PAB spacer is designed for cathepsin-mediated cleavage to release the payload under lysosomal conditions.
  • Cleavable Val-Cit-PAB spacer for enzymatic release.
  • DBCO handle for copper-free click (SPAAC) conjugation.
  • Hydrophilic PEG4 spacer improves solubility and flexibility.
  • High reported purity suitable for research conjugation workflows.
  • Available in small milligram pack sizes for lab-scale synthesis.