Learn More
Medchemexpress LLC DBCO-PEG4-Val-Cit-PAB-PNP | 2226472-28-0 | 97.3% | 1079.16 | C55H66N8O15 | 50 MG
SDP
Catalog No. 5000281941
Encompass_Preferred
DBCO-PEG4-Val-Cit-PAB-PNP is a cleavable antibody-drug conjugate (ADC) linker for bioconjugation and click-chemistry applications. It combines a dibenzocyclooctyne (DBCO) moiety for copper-free SPAAC conjugation with a PEG4 spacer for solubility and flexibility, a Val-Cit protease-cleavable dipeptide linked to a PAB self-immolative spacer, and a PNP leaving group for amine-mediated payload attachment.
  • Contains DBCO for copper-free strain-promoted azide-alkyne cycloaddition.
  • Includes PEG4 spacer to improve solubility and reduce steric hindrance.
  • Features Val-Cit dipeptide cleavable by cathepsin B for selective payload release.
  • Incorporates a PAB self-immolative spacer to enable rapid payload liberation after cleavage.
  • Provides a PNP leaving group for straightforward amine coupling of payloads.
  • Suited for synthesis of ADCs and conjugation to azide-bearing molecules.
Catalog No. 50-002-81941 Supplier Medchemexpress LLC Supplier No. HYW19091350MG
DBCO-PEG4-Val-Cit-PAB-PNP is a cleavable antibody-drug conjugate (ADC) linker for bioconjugation and click-chemistry applications. It combines a dibenzocyclooctyne (DBCO) moiety for copper-free SPAAC conjugation with a PEG4 spacer for solubility and flexibility, a Val-Cit protease-cleavable dipeptide linked to a PAB self-immolative spacer, and a PNP leaving group for amine-mediated payload attachment.
  • Contains DBCO for copper-free strain-promoted azide-alkyne cycloaddition.
  • Includes PEG4 spacer to improve solubility and reduce steric hindrance.
  • Features Val-Cit dipeptide cleavable by cathepsin B for selective payload release.
  • Incorporates a PAB self-immolative spacer to enable rapid payload liberation after cleavage.
  • Provides a PNP leaving group for straightforward amine coupling of payloads.
  • Suited for synthesis of ADCs and conjugation to azide-bearing molecules.