Learn More
Medchemexpress LLC Dbco-PEG4-VC-PAB-DMEA-PNU-159682 | 2259318-56-2 | 1695.81 g/mol | C86H106N10O26 | 1 MG
SDP
Catalog No. 5000203215
Encompass_Preferred
DBCO-PEG4-VC-PAB-DMEA-PNU-159682 is an ADC linker-drug conjugate designed for antibody-drug conjugate synthesis and click-chemistry conjugation. It contains a DBCO group for strain-promoted alkyne-azide cycloaddition, a four-unit PEG spacer for solubility, a valine-citrulline cleavable dipeptide with a para-aminobenzyl self-immolative spacer, and a DMEA-modified PNU-159682 payload.
  • Facilitates SPAAC (strain-promoted alkyne-azide cycloaddition) for bioorthogonal conjugation.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Val-Cit-PAB linker enables protease-cleavable payload release.
  • Includes a potent PNU-159682-based payload for ADC research applications.
  • Supplied in small research quantities suitable for conjugation studies.
Catalog No. 50-002-03215 Supplier Medchemexpress LLC Supplier No. HY1266911MG
DBCO-PEG4-VC-PAB-DMEA-PNU-159682 is an ADC linker-drug conjugate designed for antibody-drug conjugate synthesis and click-chemistry conjugation. It contains a DBCO group for strain-promoted alkyne-azide cycloaddition, a four-unit PEG spacer for solubility, a valine-citrulline cleavable dipeptide with a para-aminobenzyl self-immolative spacer, and a DMEA-modified PNU-159682 payload.
  • Facilitates SPAAC (strain-promoted alkyne-azide cycloaddition) for bioorthogonal conjugation.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Val-Cit-PAB linker enables protease-cleavable payload release.
  • Includes a potent PNU-159682-based payload for ADC research applications.
  • Supplied in small research quantities suitable for conjugation studies.