Learn More
Medchemexpress LLC Dbco-peg5-dbco | 2363130-04-3 | MFCD28334545 | 98.0% | C50H54N4O9 | 10MG
SDP
Catalog No. 5000261472
Encompass_Preferred
DBCO-PEG5-DBCO is a bifunctional PEG-based linker bearing two dibenzocyclooctyne (DBCO) groups designed for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry and for use as a flexible spacer in bifunctional molecule assembly, such as linker-mediated conjugates and PROTAC synthesis.
  • Enables copper-free SPAAC click conjugation to azide-containing molecules.
  • Provides a PEG5 spacer to improve solubility and flexibility of conjugates.
  • Offers two reactive DBCO termini for symmetric conjugation or crosslinking.
  • High purity (98.0%) suitable for research-grade synthesis.
  • Available in small-quantity packs to support lab-scale experimentation.
Catalog No. 50-002-61472 Supplier Medchemexpress LLC Supplier No. HY14030210MG
DBCO-PEG5-DBCO is a bifunctional PEG-based linker bearing two dibenzocyclooctyne (DBCO) groups designed for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry and for use as a flexible spacer in bifunctional molecule assembly, such as linker-mediated conjugates and PROTAC synthesis.
  • Enables copper-free SPAAC click conjugation to azide-containing molecules.
  • Provides a PEG5 spacer to improve solubility and flexibility of conjugates.
  • Offers two reactive DBCO termini for symmetric conjugation or crosslinking.
  • High purity (98.0%) suitable for research-grade synthesis.
  • Available in small-quantity packs to support lab-scale experimentation.