Learn More
Medchemexpress LLC DBCO-PEG5-NHS ester | 2144395-59-3 | 95.0% | 693.74 | C36H43N3O11 | 25 MG
SDP
Catalog No. 5000202330
Encompass_Preferred
DBCO-PEG5-NHS ester is a DBCO-functionalized PEG linker with an N-hydroxysuccinimide (NHS) ester reactive group. It acts as a click chemistry reagent enabling strain-promoted alkyne-azide cycloaddition (SPAAC) and serves as a PEG-based linker for the synthesis of PROTACs and antibody-drug conjugates (ADCs).
  • Enables copper-free click chemistry (SPAAC) with azide-containing molecules.
  • Reactive NHS ester couples efficiently to primary amines, such as lysine residues or aminosilane-coated surfaces.
  • PEG5 spacer provides flexibility and reduces steric hindrance at conjugation sites.
  • Suitable for linker design in PROTAC and ADC synthesis.
  • Available in small milligram pack sizes for research-scale applications.
Catalog No. 50-002-02330 Supplier Medchemexpress LLC Supplier No. HY12688525MG
DBCO-PEG5-NHS ester is a DBCO-functionalized PEG linker with an N-hydroxysuccinimide (NHS) ester reactive group. It acts as a click chemistry reagent enabling strain-promoted alkyne-azide cycloaddition (SPAAC) and serves as a PEG-based linker for the synthesis of PROTACs and antibody-drug conjugates (ADCs).
  • Enables copper-free click chemistry (SPAAC) with azide-containing molecules.
  • Reactive NHS ester couples efficiently to primary amines, such as lysine residues or aminosilane-coated surfaces.
  • PEG5 spacer provides flexibility and reduces steric hindrance at conjugation sites.
  • Suitable for linker design in PROTAC and ADC synthesis.
  • Available in small milligram pack sizes for research-scale applications.