Learn More
Medchemexpress LLC DBCO-PEG6-NHS ester | 95.0% | 737.79 | 50 MG
SDP
Catalog No. 5000391992
Encompass_Preferred
DBCO-PEG6-NHS ester is a click chemistry PEG reagent containing an NHS ester that reacts specifically with primary amines, such as side chains of lysine residues or aminosilane-coated surfaces, under neutral or slightly alkaline conditions. It reacts efficiently to form covalent bonds. DBCO-PEG6-NHS ester also contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • Contains an NHS ester that reacts specifically with primary amines.
  • Reacts under neutral or slightly alkaline conditions.
  • Forms covalent bonds.
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
Catalog No. 50-003-91992 Supplier Medchemexpress LLC Supplier No. HY16077350MG
DBCO-PEG6-NHS ester is a click chemistry PEG reagent containing an NHS ester that reacts specifically with primary amines, such as side chains of lysine residues or aminosilane-coated surfaces, under neutral or slightly alkaline conditions. It reacts efficiently to form covalent bonds. DBCO-PEG6-NHS ester also contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • Contains an NHS ester that reacts specifically with primary amines.
  • Reacts under neutral or slightly alkaline conditions.
  • Forms covalent bonds.
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.