Learn More
Medchemexpress LLC DBCO-PEG6-NHS ester | 95.0% | 737.79 g·mol⁻¹ | C38H47N3O12 | 10 MG
SDP
Catalog No. 5000285412
Encompass_Preferred
DBCO-PEG6-NHS ester is a bifunctional click-chemistry reagent combining a dibenzocyclooctyne (DBCO) moiety and an N-hydroxysuccinimide (NHS) ester separated by a six-unit polyethylene glycol spacer. It is designed for amine-specific conjugation and copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners, and is supplied as a light yellow liquid.
  • Bifunctional linker with DBCO and NHS ester groups.
  • PEG6 spacer improves solubility and reduces steric hindrance.
  • Reacts with primary amines for stable covalent attachment.
  • Performs copper-free SPAAC with azide groups for bioorthogonal labeling.
  • Purity approximately 95% as provided in product documentation.
  • Store protected from light and under inert atmosphere; recommended low-temperature storage for long-term stability.
Catalog No. 50-002-85412 Supplier Medchemexpress LLC Supplier No. HY16077310MG
DBCO-PEG6-NHS ester is a bifunctional click-chemistry reagent combining a dibenzocyclooctyne (DBCO) moiety and an N-hydroxysuccinimide (NHS) ester separated by a six-unit polyethylene glycol spacer. It is designed for amine-specific conjugation and copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners, and is supplied as a light yellow liquid.
  • Bifunctional linker with DBCO and NHS ester groups.
  • PEG6 spacer improves solubility and reduces steric hindrance.
  • Reacts with primary amines for stable covalent attachment.
  • Performs copper-free SPAAC with azide groups for bioorthogonal labeling.
  • Purity approximately 95% as provided in product documentation.
  • Store protected from light and under inert atmosphere; recommended low-temperature storage for long-term stability.