Learn More
Medchemexpress LLC DBCO-PEG6-NHS ester | 95.99% | 737.79 | 25 MG
SDP
Catalog No. 5000392033
Encompass_Preferred
DBCO-PEG6-NHS ester is a click chemistry PEG reagent containing an NHS ester that reacts specifically with primary amines, such as side chains of lysine residues or aminosilane-coated surfaces, under neutral or slightly alkaline conditions, to form covalent bonds. It also contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • Reacts specifically with primary amines via NHS ester
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
  • Suitable for covalent bond formation
  • Useful in click chemistry reactions
Catalog No. 50-003-92033 Supplier Medchemexpress LLC Supplier No. 25MGHY160773
DBCO-PEG6-NHS ester is a click chemistry PEG reagent containing an NHS ester that reacts specifically with primary amines, such as side chains of lysine residues or aminosilane-coated surfaces, under neutral or slightly alkaline conditions, to form covalent bonds. It also contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • Reacts specifically with primary amines via NHS ester
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
  • Suitable for covalent bond formation
  • Useful in click chemistry reactions