Learn More
Medchemexpress LLC DBCO-PEG8-NHS ester | 2553412-88-5 | 98.3% | C42H55N3O14 | 25 MG
SDP
Catalog No. 5000355936
Encompass_Preferred
DBCO-PEG8-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent features a DBCO group, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs selectively degrade target proteins by utilizing the intracellular ubiquitin-proteasome system.
  • Functions as a click chemistry reagent
  • Undergoes strain-promoted alkyne-azide cycloaddition
  • Utilizes the ubiquitin-proteasome system for protein degradation
  • Suitable for PROTAC synthesis
Catalog No. 50-003-55936 Supplier Medchemexpress LLC Supplier No. 25MGHY140273
DBCO-PEG8-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent features a DBCO group, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs selectively degrade target proteins by utilizing the intracellular ubiquitin-proteasome system.
  • Functions as a click chemistry reagent
  • Undergoes strain-promoted alkyne-azide cycloaddition
  • Utilizes the ubiquitin-proteasome system for protein degradation
  • Suitable for PROTAC synthesis