Learn More
Medchemexpress LLC DBCO-PEG8-NHS ester | 2553412-88-5 | 98.3% | C42H55N3O14 | 50 MG
SDP
Catalog No. 5000353834
Encompass_Preferred
DBCO-PEG8-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that enables strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing azide groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Features a DBCO group for SPAAC with azide-containing molecules
  • Used in the synthesis of PROTACs
Catalog No. 50-003-53834 Supplier Medchemexpress LLC Supplier No. HY14027350MG
DBCO-PEG8-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that enables strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing azide groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Features a DBCO group for SPAAC with azide-containing molecules
  • Used in the synthesis of PROTACs