Learn More
Broadpharm DBCO-PEG8-NHS ester, 50mg
Catalog No. NC1617311
Encompass
Formula: C42H55N3O14, Purity: 98%, MW: 825.9, DBCO-PEG8-NHS Ester is a PEG derivative containing an NHS ester moiety that can react specifically and efficiently with primary amines (e.g. the side chain of lysine residues or aminosilane-coated surfaces) at neutral or slightly basic condition to form a covalent bond. The hydrophilic PEG spacer arm improves water solubility and provides a long and flexible connection that minimizes steric hindrance involved with ligation. DBCO is commonly used for copper-free Click Chemistry reactions.
Catalog No. NC1617311 Supplier Broadpharm Supplier No. BP2401950MG
Formula: C42H55N3O14, Purity: 98%, MW: 825.9, DBCO-PEG8-NHS Ester is a PEG derivative containing an NHS ester moiety that can react specifically and efficiently with primary amines (e.g. the side chain of lysine residues or aminosilane-coated surfaces) at neutral or slightly basic condition to form a covalent bond. The hydrophilic PEG spacer arm improves water solubility and provides a long and flexible connection that minimizes steric hindrance involved with ligation. DBCO is commonly used for copper-free Click Chemistry reactions.