Learn More
Medchemexpress LLC DBCO-PEG9-amine | 2353409-99-9 | 95.0% | C39H57N3O11 | 25 MG
SDP
Catalog No. 5000357394
Encompass_Preferred
DBCO-PEG9-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing Azide groups. PROTACs themselves consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation. They operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a DBCO group for SPAAC reactions
  • Facilitates strain-promoted alkyne-azide cycloaddition with azide groups
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57394 Supplier Medchemexpress LLC Supplier No. 25MGHY140285
DBCO-PEG9-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing Azide groups. PROTACs themselves consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation. They operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a DBCO group for SPAAC reactions
  • Facilitates strain-promoted alkyne-azide cycloaddition with azide groups
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins