Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-PEG9-amine | 2353409-99-9 | C39H57N3O11 | 50 MG
SDP

Catalog No. 5000353980
Encompass_Preferred

DBCO-PEG9-amine is a PEG-based PROTAC linker designed for use in PROTAC synthesis. This click chemistry reagent features a DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.

  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains a DBCO group for SPAAC reactions
  • PEG-based linker structure
  • Molecular weight: 743.88
  • Appearance: light yellow to yellow oil
  • Recommended storage at -20°C, protected from light and under argon
  • Soluble in DMSO up to 100 mg/mL

Catalog No. 50-003-53980 Supplier Medchemexpress LLC Supplier No. HY14028550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG9-amine is a PEG-based PROTAC linker designed for use in PROTAC synthesis. This click chemistry reagent features a DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.

  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains a DBCO group for SPAAC reactions
  • PEG-based linker structure
  • Molecular weight: 743.88
  • Appearance: light yellow to yellow oil
  • Recommended storage at -20°C, protected from light and under argon
  • Soluble in DMSO up to 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.