Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbco-s-s-peg3-biotin | 1430408-09-5 | MFCD28334542 | 96.9% | 869.12 g/mol | C42H56N6O8S3 | 5 MG
SDP

Catalog No. 5000223248
Encompass_Preferred

DBCO-S-S-PEG3-biotin is a PEG-based cleavable biotinylation reagent for copper-free click chemistry and PROTAC linker synthesis. It combines a DBCO reactive moiety with a disulfide-containing PEG3 spacer to enable strain-promoted alkyne-azide cycloaddition and reductive release of the biotin tag.

  • Contains a DBCO group enabling copper-free strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Includes a disulfide cleavable linker for release under reducing conditions.
  • Features a PEG3 spacer to improve solubility and reduce steric hindrance.
  • Provides a biotin tag for affinity capture and purification workflows.
  • Soluble in DMSO at high concentration and compatible with common formulation protocols.
  • Supplied as a colorless to off-white solid with high purity.

Catalog No. 50-002-23248 Supplier Medchemexpress LLC Supplier No. HY1401285MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-S-S-PEG3-biotin is a PEG-based cleavable biotinylation reagent for copper-free click chemistry and PROTAC linker synthesis. It combines a DBCO reactive moiety with a disulfide-containing PEG3 spacer to enable strain-promoted alkyne-azide cycloaddition and reductive release of the biotin tag.

  • Contains a DBCO group enabling copper-free strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Includes a disulfide cleavable linker for release under reducing conditions.
  • Features a PEG3 spacer to improve solubility and reduce steric hindrance.
  • Provides a biotin tag for affinity capture and purification workflows.
  • Soluble in DMSO at high concentration and compatible with common formulation protocols.
  • Supplied as a colorless to off-white solid with high purity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.