Learn More
Medchemexpress LLC DBCO-Sulfo-Link-biot 1mg
SDP
Catalog No. 5000206591
Encompass_Preferred
DBCO-Sulfo-Link-biotin TEA is a click chemistry intermediate containing a DBCO group which can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing an azide group
Catalog No. 50-002-06591 Supplier Medchemexpress LLC Supplier No. HYW1230191MG
DBCO-Sulfo-Link-biotin TEA is a click chemistry intermediate containing a DBCO group which can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing an azide group