Learn More
Medchemexpress LLC DBCO-Sulfo-Link-biot 5mg
SDP
Catalog No. 5000206776
Encompass_Preferred
DBCO-Sulfo-Link-biotin TEA is a click chemistry intermediate containing a DBCO group which can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing an azide group
Catalog No. 50-002-06776 Supplier Medchemexpress LLC Supplier No. HYW1230195MG
DBCO-Sulfo-Link-biotin TEA is a click chemistry intermediate containing a DBCO group which can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing an azide group