Learn More
Medchemexpress LLC Dbco-sulfo-link-biotin | 1363444-70-5 | 653.77 g·mol⁻¹ | C31H35N5O7S2 | 1 MG
SDP
Catalog No. 5000204260
Encompass_Preferred
DBCO-Sulfo-Link-biotin is a water-soluble, cleavable click-chemistry linker that combines a dibenzocyclooctyne (DBCO) reactive group with a biotin affinity tag. It enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) for labeling azide-containing biomolecules and is used in biotinylation, protein labeling, and antibody-drug conjugate linker synthesis.
  • Enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Provides a biotin tag for affinity capture and detection.
  • Sulfo linker increases water solubility for aqueous bioconjugation.
  • Suitable for biotinylation, protein labeling, and ADC linker incorporation.
Catalog No. 50-002-04260 Supplier Medchemexpress LLC Supplier No. HY1308101MG
DBCO-Sulfo-Link-biotin is a water-soluble, cleavable click-chemistry linker that combines a dibenzocyclooctyne (DBCO) reactive group with a biotin affinity tag. It enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) for labeling azide-containing biomolecules and is used in biotinylation, protein labeling, and antibody-drug conjugate linker synthesis.
  • Enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Provides a biotin tag for affinity capture and detection.
  • Sulfo linker increases water solubility for aqueous bioconjugation.
  • Suitable for biotinylation, protein labeling, and ADC linker incorporation.